Assay Details
| Gene Symbol: svr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144669.2 | NP_001138141.1 | 4 - 5 | 1833 | 54 | |
| NM_001297815.1 | NP_001284744.1 | 4 - 5 | 1467 | 54 | ||
| NM_080293.4 | NP_525032.2 | 5 - 6 | 1746 | 54 | ||
| NM_166846.5 | NP_726675.3 | 4 - 5 | 1760 | 54 | ||
| NM_176679.3 | NP_788852.2 | 4 - 5 | 1467 | 54 | ||
| NM_206596.2 | NP_996319.1 | 5 - 6 | 1746 | 54 | ||
| NM_206599.2 | NP_996322.1 | 4 - 5 | 1760 | 54 | ||
| GenBank mRNA | AF545816.1 | - | 4 - 5 | 1404 | 54 | |
| AF545817.1 | - | 4 - 5 | 1524 | 54 | ||
| AF545818.1 | - | 4 - 5 | 1524 | 54 | ||
| BT099720.1 | - | 4 - 5 | 1437 | 54 | ||
| BT100310.1 | - | 4 - 5 | 946 | 54 | ||
More Information
Set Membership: |
|

svr
Genomic Map