Assay Details
| Gene Symbol: Gclc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298072.1 | NP_001285001.1 | 3 - 4 | 2516 | 58 | |
| NM_001298073.1 | NP_001285002.1 | 3 - 4 | 1910 | 58 | ||
| NM_080266.3 | NP_525005.2 | 4 - 5 | 2105 | 58 | ||
| NM_206650.2 | NP_996373.1 | 3 - 4 | 1824 | 58 | ||
| GenBank mRNA | AY071586.1 | - | 4 - 5 | 2102 | 58 | |
More Information
Set Membership: |
|

Gclc
Genomic Map