Assay Details
| Gene Symbol: otu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272403.1 | NP_001259332.1 | 4 - 5 | 551 | 125 | |
| NM_001272404.1 | NP_001259333.1 | 4 - 5 | 663 | 125 | ||
| NM_078534.4 | NP_511089.2 | 4 - 5 | 646 | 125 | ||
| NM_167158.3 | NP_727271.1 | 4 - 5 | 646 | 125 | ||
| NM_206656.2 | NP_996379.1 | 4 - 5 | 660 | 125 | ||
| GenBank mRNA | BT133210.1 | - | 4 - 5 | 611 | 125 | |
More Information
Set Membership: |
|

otu
Genomic Map