Assay Details
| Gene Symbol: otu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078534.4 | NP_511089.2 | 7 - 8 | 1311 | 87 | |
| NM_206656.2 | NP_996379.1 | 7 - 8 | 1325 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

otu| Gene Symbol: otu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078534.4 | NP_511089.2 | 7 - 8 | 1311 | 87 | |
| NM_206656.2 | NP_996379.1 | 7 - 8 | 1325 | 87 | ||
Set Membership: |
|