Assay Details
| Gene Symbol: spri | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103471.2 | NP_001096941.2 | 4 - 5 | 2253 | 73 | |
| NM_001272484.1 | NP_001259413.1 | 6 - 7 | 3134 | 73 | ||
| NM_001272485.1 | NP_001259414.1 | 6 - 7 | 3219 | 73 | ||
| NM_001272486.1 | NP_001259415.1 | 7 - 8 | 3601 | 73 | ||
| NM_001272487.1 | NP_001259416.1 | 7 - 8 | 3523 | 73 | ||
| NM_001272488.1 | NP_001259417.1 | 4 - 5 | 2384 | 73 | ||
| NM_001272489.1 | NP_001259418.1 | 7 - 8 | 4537 | 73 | ||
| NM_001272492.1 | NP_001259421.1 | 5 - 6 | 3821 | 73 | ||
| NM_176723.3 | NP_788896.1 | 6 - 7 | 3222 | 73 | ||
| NM_206678.2 | NP_996401.1 | 6 - 7 | 3775 | 73 | ||
More Information
Set Membership: |
|

spri
Genomic Map