Assay Details
| Gene Symbol: slgA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258863.2 | NP_001245792.1 | 2 - 3 | 668 | 78 | |
| NM_001258864.2 | NP_001245793.1 | 2 - 3 | 600 | 78 | ||
| NM_001258865.2 | NP_001245794.1 | 1 - 2 | 555 | 78 | ||
| NM_001258867.2 | NP_001245796.1 | 2 - 3 | 617 | 78 | ||
| NM_078709.4 | NP_523433.2 | 2 - 3 | 974 | 78 | ||
| NM_167751.3 | NP_728422.1 | 2 - 3 | 974 | 78 | ||
| NM_206803.3 | NP_996526.1 | 2 - 3 | 1055 | 78 | ||
| NM_206804.3 | NP_996527.1 | 2 - 3 | 617 | 78 | ||
| NM_206805.3 | NP_996528.1 | 2 - 3 | 693 | 78 | ||
| GenBank mRNA | AY069407.1 | - | 2 - 3 | 945 | 78 | |
More Information
Set Membership: |
|

slgA
Genomic Map