Assay Details
| Gene Symbol: slgA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258862.2 | NP_001245791.1 | 9 - 10 | 1595 | 64 | |
| NM_001258863.2 | NP_001245792.1 | 8 - 9 | 1487 | 64 | ||
| NM_001258864.2 | NP_001245793.1 | 8 - 9 | 1419 | 64 | ||
| NM_001258865.2 | NP_001245794.1 | 7 - 8 | 1374 | 64 | ||
| NM_001258866.2 | NP_001245795.1 | 8 - 9 | 1436 | 64 | ||
| NM_001258867.2 | NP_001245796.1 | 9 - 10 | 1472 | 64 | ||
| NM_078709.4 | NP_523433.2 | 9 - 10 | 1829 | 64 | ||
| NM_167751.3 | NP_728422.1 | 8 - 9 | 1793 | 64 | ||
| NM_167752.3 | NP_728423.1 | 9 - 10 | 1829 | 64 | ||
| NM_167753.3 | NP_728424.1 | 8 - 9 | 1793 | 64 | ||
| NM_167754.2 | NP_728425.1 | 3 - 4 | 509 | 64 | ||
| NM_206803.3 | NP_996526.1 | 8 - 9 | 1874 | 64 | ||
| NM_206804.3 | NP_996527.1 | 8 - 9 | 1436 | 64 | ||
| NM_206805.3 | NP_996528.1 | 8 - 9 | 1512 | 64 | ||
| GenBank mRNA | AY069407.1 | - | 8 - 9 | 1764 | 64 | |
More Information
Set Membership: |
|

slgA
Genomic Map