Assay Details
| Gene Symbol: mRpL48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272949.1 | NP_001259878.1 | 2 - 3 | 178 | 62 | |
| NM_134769.3 | NP_608613.1 | 2 - 3 | 178 | 62 | ||
| GenBank mRNA | AY069759.1 | - | 2 - 3 | 173 | 62 | |
More Information
Set Membership: |
|

mRpL48
Genomic Map