Assay Details
| Gene Symbol: CG17646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134774.3 | NP_608618.1 | 4 - 5 | 1289 | 78 | |
| NM_164436.2 | NP_722727.1 | 4 - 5 | 867 | 78 | ||
More Information
Set Membership: |
|

CG17646
Genomic Map