Assay Details
| Gene Symbol: Dim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134986.3 | NP_608830.3 | 1 - 2 | 165 | 104 | |
| GenBank mRNA | AY071048.1 | - | 1 - 2 | 161 | 104 | |
More Information
Set Membership: |
|

Dim1
Genomic Map