Assay Details
| Gene Symbol: Doc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274683.1 | NP_001261612.1 | 3 - 4 | 500 | 86 | |
| NM_140026.3 | NP_648283.1 | 3 - 4 | 687 | 86 | ||
| GenBank mRNA | AB035412.1 | - | 3 - 4 | 678 | 86 | |
| BT025226.1 | - | 3 - 4 | 685 | 86 | ||
More Information
Set Membership: |
|

Doc1
Genomic Map