Assay Details
| Gene Symbol: Mob2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274784.1 | NP_001261713.1 | 3 - 4 | 655 | 73 | |
| NM_140217.2 | NP_648474.1 | 2 - 3 | 495 | 73 | ||
| NM_168460.2 | NP_729714.1 | 2 - 3 | 623 | 73 | ||
| NM_168461.2 | NP_729715.2 | 3 - 4 | 1495 | 73 | ||
| NM_168462.2 | NP_729716.1 | 3 - 4 | 606 | 73 | ||
| GenBank mRNA | BT124999.1 | - | 3 - 4 | 586 | 73 | |
More Information
Set Membership: |
|

Mob2
Genomic Map