Assay Details
| Gene Symbol: Sms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300115.1 | NP_001287044.1 | 1 - 2 | 361 | 83 | |
| NM_140308.3 | NP_648565.1 | 1 - 2 | 361 | 83 | ||
| GenBank mRNA | AY071085.1 | - | 1 - 2 | 376 | 83 | |
| BT003439.1 | - | 1 - 2 | 377 | 83 | ||
More Information
Set Membership: |
|

Sms
Genomic Map