Assay Details
| Gene Symbol: Sms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300115.1 | NP_001287044.1 | 3 - 4 | 589 | 74 | |
| NM_001300116.1 | NP_001287045.1 | 3 - 4 | 592 | 74 | ||
| NM_140308.3 | NP_648565.1 | 3 - 4 | 589 | 74 | ||
| NM_168496.2 | NP_729798.1 | 3 - 4 | 592 | 74 | ||
| GenBank mRNA | AY071085.1 | - | 3 - 4 | 604 | 74 | |
| BT001432.1 | - | 3 - 4 | 567 | 74 | ||
| BT003439.1 | - | 3 - 4 | 605 | 74 | ||
More Information
Set Membership: |
|

Sms
Genomic Map