Assay Details
| Gene Symbol: sti | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274839.1 | NP_001261768.1 | 5 - 6 | 3917 | 96 | |
| NM_140327.2 | NP_648584.1 | 5 - 6 | 3905 | 96 | ||
| GenBank mRNA | BT081968.1 | - | 5 - 6 | 3859 | 96 | |
More Information
Set Membership: |
|

sti
Genomic Map