Assay Details
| Gene Symbol: sti | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274839.1 | NP_001261768.1 | 6 - 7 | 4110 | 90 | |
| NM_140327.2 | NP_648584.1 | 6 - 7 | 4098 | 90 | ||
| GenBank mRNA | BT081968.1 | - | 6 - 7 | 4052 | 90 | |
More Information
Set Membership: |
|

sti
Genomic Map