Assay Details
| Gene Symbol: Atg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169962.2 | NP_001163433.1 | 7 - 8 | 1729 | 88 | |
| NM_140344.3 | NP_648601.1 | 8 - 9 | 1669 | 88 | ||
| GenBank mRNA | AY129445.1 | - | 7 - 8 | 1698 | 88 | |
| BT021340.1 | - | 8 - 9 | 1604 | 88 | ||
More Information
Set Membership: |
|

Atg1
Genomic Map