Assay Details
| Gene Symbol: Nhe3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103635.3 | NP_001097105.3 | 2 - 3 | 766 | 59 | |
| NM_001144329.2 | NP_001137801.1 | 2 - 3 | 759 | 59 | ||
| NM_001273217.1 | NP_001260146.1 | 2 - 3 | 259 | 59 | ||
| NM_001273218.1 | NP_001260147.1 | 2 - 3 | 759 | 59 | ||
| NM_135220.3 | NP_609064.3 | 2 - 3 | 759 | 59 | ||
| NM_164701.2 | NP_723202.2 | 2 - 3 | 759 | 59 | ||
| GenBank mRNA | BT124889.1 | - | 2 - 3 | 259 | 59 | |
More Information
Set Membership: |
|

Nhe3
Genomic Map