Assay Details
| Gene Symbol: AkhR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273220.1 | NP_001260149.1 | 2 - 3 | 930 | 75 | |
| NM_058039.4 | NP_477387.1 | 2 - 3 | 374 | 75 | ||
| NM_164705.3 | NP_723206.1 | 3 - 4 | 529 | 75 | ||
| NM_205917.3 | NP_995639.1 | 2 - 3 | 374 | 75 | ||
| GenBank mRNA | BT031198.1 | - | 2 - 3 | 354 | 75 | |
More Information
Set Membership: |
|

AkhR
Genomic Map