Assay Details
| Gene Symbol: AkhR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273220.1 | NP_001260149.1 | 3 - 4 | 1198 | 62 | |
| NM_058039.4 | NP_477387.1 | 3 - 4 | 642 | 62 | ||
| NM_164705.3 | NP_723206.1 | 4 - 5 | 797 | 62 | ||
| NM_205917.3 | NP_995639.1 | 3 - 4 | 642 | 62 | ||
| GenBank mRNA | BT031198.1 | - | 3 - 4 | 622 | 62 | |
More Information
Set Membership: |
|

AkhR
Genomic Map