Assay Details
| Gene Symbol: Ntl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164765.4 | NP_723303.1 | 1 - 2 | 118 | 71 | |
| GenBank mRNA | BT133486.1 | - | 1 - 2 | 115 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ntl| Gene Symbol: Ntl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164765.4 | NP_723303.1 | 1 - 2 | 118 | 71 | |
| GenBank mRNA | BT133486.1 | - | 1 - 2 | 115 | 71 | |
Set Membership: |
|