Assay Details
| Gene Symbol: Ntl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135291.3 | NP_609135.1 | 3 - 4 | 676 | 102 | |
| NM_164765.4 | NP_723303.1 | 3 - 4 | 467 | 102 | ||
| GenBank mRNA | BT133486.1 | - | 3 - 4 | 464 | 102 | |
More Information
Set Membership: |
|

Ntl
Genomic Map