Assay Details
| Gene Symbol: Btk29A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103650.2 | NP_001097120.1 | 7 - 8 | 3727 | 62 | |
| NM_001103651.3 | NP_001097121.1 | 11 - 12 | 2388 | 62 | ||
| NM_001273314.1 | NP_001260243.1 | 7 - 8 | 2037 | 62 | ||
| NM_001273315.1 | NP_001260244.1 | 10 - 11 | 2561 | 62 | ||
| NM_057397.6 | NP_476745.1 | 11 - 12 | 2721 | 62 | ||
| NM_057398.4 | NP_476746.1 | 8 - 9 | 2011 | 62 | ||
| NM_164804.2 | NP_723369.1 | 11 - 12 | 2409 | 62 | ||
| NM_164805.2 | NP_723370.1 | 8 - 9 | 2014 | 62 | ||
| GenBank mRNA | AY069457.1 | - | 7 - 8 | 3858 | 62 | |
| AY128441.1 | - | 11 - 12 | 2676 | 62 | ||
More Information
Set Membership: |
|

Btk29A
Genomic Map