Assay Details
| Gene Symbol: Btk29A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103651.3 | NP_001097121.1 | 4 - 5 | 799 | 77 | |
| NM_001273315.1 | NP_001260244.1 | 3 - 4 | 972 | 77 | ||
| NM_057397.6 | NP_476745.1 | 4 - 5 | 1132 | 77 | ||
| NM_164804.2 | NP_723369.1 | 4 - 5 | 820 | 77 | ||
| GenBank mRNA | AY128441.1 | - | 4 - 5 | 1087 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001273315.1 transcripts.
Set Membership: |
|

Btk29A
Genomic Map