Assay Details
| Gene Symbol: Btk29A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103650.2 | NP_001097120.1 | 2 - 3 | 2493 | 81 | |
| NM_001103651.3 | NP_001097121.1 | 6 - 7 | 1154 | 81 | ||
| NM_001273314.1 | NP_001260243.1 | 2 - 3 | 803 | 81 | ||
| NM_001273315.1 | NP_001260244.1 | 5 - 6 | 1327 | 81 | ||
| NM_057397.6 | NP_476745.1 | 6 - 7 | 1487 | 81 | ||
| NM_057398.4 | NP_476746.1 | 3 - 4 | 777 | 81 | ||
| NM_164804.2 | NP_723369.1 | 6 - 7 | 1175 | 81 | ||
| NM_164805.2 | NP_723370.1 | 3 - 4 | 780 | 81 | ||
| GenBank mRNA | AY069457.1 | - | 2 - 3 | 2624 | 81 | |
| AY128441.1 | - | 6 - 7 | 1442 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001273315.1 transcripts.
Set Membership: |
|

Btk29A
Genomic Map