Assay Details
| Gene Symbol: Btk29A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103650.2 | NP_001097120.1 | 5 - 6 | 2829 | 94 | |
| NM_001103651.3 | NP_001097121.1 | 9 - 10 | 1490 | 94 | ||
| NM_001273314.1 | NP_001260243.1 | 5 - 6 | 1139 | 94 | ||
| NM_001273315.1 | NP_001260244.1 | 8 - 9 | 1663 | 94 | ||
| NM_057397.6 | NP_476745.1 | 9 - 10 | 1823 | 94 | ||
| NM_057398.4 | NP_476746.1 | 6 - 7 | 1113 | 94 | ||
| NM_164804.2 | NP_723369.1 | 9 - 10 | 1511 | 94 | ||
| NM_164805.2 | NP_723370.1 | 6 - 7 | 1116 | 94 | ||
| GenBank mRNA | AY069457.1 | - | 5 - 6 | 2960 | 94 | |
| AY128441.1 | - | 9 - 10 | 1778 | 94 | ||
More Information
Set Membership: |
|

Btk29A
Genomic Map