Assay Details
| Gene Symbol: Btk29A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103650.2 | NP_001097120.1 | 1 - 2 | 2292 | 66 | |
| NM_057398.4 | NP_476746.1 | 2 - 3 | 576 | 66 | ||
| NM_164805.2 | NP_723370.1 | 2 - 3 | 579 | 66 | ||
| GenBank mRNA | AY069457.1 | - | 1 - 2 | 2423 | 66 | |
More Information
Set Membership: |
|

Btk29A
Genomic Map