Assay Details
| Gene Symbol: Sema1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201802.2 | NP_001188731.1 | 15 - 16 | 2833 | 68 | |
| NM_001201803.2 | NP_001188732.1 | 14 - 15 | 2947 | 68 | ||
| NM_001273336.1 | NP_001260265.1 | 14 - 15 | 2947 | 68 | ||
| NM_001273337.1 | NP_001260266.1 | 14 - 15 | 2906 | 68 | ||
| NM_001273338.1 | NP_001260267.1 | 14 - 15 | 2947 | 68 | ||
| NM_135399.5 | NP_609243.3 | 14 - 15 | 2627 | 68 | ||
| NM_164830.5 | NP_723407.2 | 14 - 15 | 2947 | 68 | ||
More Information
Set Membership: |
|

Sema1a
Genomic Map