Assay Details
| Gene Symbol: Sema1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273337.1 | NP_001260266.1 | 17 - 18 | 3294 | 88 | |
| NM_001273338.1 | NP_001260267.1 | 17 - 18 | 3335 | 88 | ||
| NM_164830.5 | NP_723407.2 | 17 - 18 | 3335 | 88 | ||
More Information
Set Membership: |
|

Sema1a
Genomic Map