Assay Details
| Gene Symbol: RpL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298897.1 | NP_001285826.1 | 3 - 4 | 328 | 64 | |
| NM_057813.4 | NP_477161.1 | 3 - 4 | 364 | 64 | ||
| NM_164955.2 | NP_723644.1 | 2 - 3 | 401 | 64 | ||
| GenBank mRNA | BT022718.1 | - | 3 - 4 | 364 | 64 | |
More Information
Set Membership: |
|

RpL9
Genomic Map