Assay Details
| Gene Symbol: Gmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135044.4 | NP_608888.2 | 1 - 2 | 261 | 87 | |
| GenBank mRNA | BT099628.1 | - | 1 - 2 | 227 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gmd| Gene Symbol: Gmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135044.4 | NP_608888.2 | 1 - 2 | 261 | 87 | |
| GenBank mRNA | BT099628.1 | - | 1 - 2 | 227 | 87 | |
Set Membership: |
|