Assay Details
| Gene Symbol: Reck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274924.1 | NP_001261853.1 | 2 - 3 | 756 | 73 | |
| NM_140476.3 | NP_648733.2 | 2 - 3 | 788 | 73 | ||
| GenBank mRNA | BT150289.1 | - | 2 - 3 | 756 | 73 | |
More Information
Set Membership: |
|

Reck
Genomic Map