Assay Details
| Gene Symbol: arx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140814.2 | NP_649071.1 | 1 - 2 | 232 | 90 | |
| GenBank mRNA | AY095030.1 | - | 1 - 2 | 171 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

arx| Gene Symbol: arx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140814.2 | NP_649071.1 | 1 - 2 | 232 | 90 | |
| GenBank mRNA | AY095030.1 | - | 1 - 2 | 171 | 90 | |
Set Membership: |
|