Assay Details
| Gene Symbol: Mtr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140900.5 | NP_649157.1 | 1 - 2 | 234 | 72 | |
| GenBank mRNA | BT030767.1 | - | 1 - 2 | 214 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtr3| Gene Symbol: Mtr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140900.5 | NP_649157.1 | 1 - 2 | 234 | 72 | |
| GenBank mRNA | BT030767.1 | - | 1 - 2 | 214 | 72 | |
Set Membership: |
|