Assay Details
| Gene Symbol: MRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165020.3 | NP_723772.2 | 7 - 8 | 3282 | 75 | |
| NM_205971.2 | NP_995693.1 | 7 - 8 | 3278 | 75 | ||
| NM_205980.2 | NP_995702.1 | 8 - 9 | 3396 | 75 | ||
More Information
Set Membership: |
|

MRP
Genomic Map