Assay Details
| Gene Symbol: mnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014750.2 | NP_001014750.1 | 5 - 6 | 2367 | 110 | |
| NM_001298464.1 | NP_001285393.1 | 4 - 5 | 1088 | 110 | ||
| NM_167581.3 | NP_728104.1 | 5 - 6 | 2367 | 110 | ||
| NM_170658.3 | NP_728106.1 | 5 - 6 | 2367 | 110 | ||
| NM_170659.2 | NP_728107.2 | 4 - 5 | 1088 | 110 | ||
More Information
Set Membership: |
|

mnb
Genomic Map