Assay Details
| Gene Symbol: Sam-S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258885.2 | NP_001245814.1 | 3 - 4 | 377 | 67 | |
| NM_001272862.1 | NP_001259791.1 | 3 - 4 | 495 | 67 | ||
| NM_080184.2 | NP_524923.1 | 3 - 4 | 464 | 67 | ||
| NM_164355.3 | NP_722593.1 | 3 - 4 | 393 | 67 | ||
| NM_164356.2 | NP_722594.1 | 3 - 4 | 377 | 67 | ||
| NM_164357.2 | NP_722595.1 | 3 - 4 | 464 | 67 | ||
| NM_164358.2 | NP_722596.1 | 3 - 4 | 437 | 67 | ||
| NM_164359.2 | NP_722597.1 | 3 - 4 | 589 | 67 | ||
| NM_164360.3 | NP_722598.1 | 3 - 4 | 513 | 67 | ||
| NM_164361.2 | NP_722599.1 | 3 - 4 | 377 | 67 | ||
| NM_164362.3 | NP_722600.1 | 2 - 3 | 277 | 67 | ||
| NM_205880.1 | NP_995602.1 | 4 - 5 | 471 | 67 | ||
| GenBank mRNA | AY051918.1 | - | 3 - 4 | 373 | 67 | |
| BT099838.1 | - | 2 - 3 | 263 | 67 | ||
| BT125773.1 | - | 3 - 4 | 354 | 67 | ||
More Information
Set Membership: |
|

Sam-S
Genomic Map