Assay Details
| Gene Symbol: Sam-S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164356.2 | NP_722594.1 | 5 - 6 | 617 | 65 | |
| NM_164357.2 | NP_722595.1 | 5 - 6 | 704 | 65 | ||
| NM_164358.2 | NP_722596.1 | 5 - 6 | 677 | 65 | ||
| NM_164359.2 | NP_722597.1 | 5 - 6 | 829 | 65 | ||
| NM_164360.3 | NP_722598.1 | 5 - 6 | 753 | 65 | ||
| NM_164361.2 | NP_722599.1 | 6 - 7 | 730 | 65 | ||
| NM_205880.1 | NP_995602.1 | 6 - 7 | 711 | 65 | ||
| GenBank mRNA | AY051918.1 | - | 5 - 6 | 613 | 65 | |
More Information
Set Membership: |
|

Sam-S
Genomic Map