Assay Details
| Gene Symbol: Sam-S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272862.1 | NP_001259791.1 | 5 - 6 | 1299 | 132 | |
| NM_080184.2 | NP_524923.1 | 6 - 7 | 1381 | 132 | ||
| NM_164355.3 | NP_722593.1 | 6 - 7 | 1310 | 132 | ||
| NM_164356.2 | NP_722594.1 | 6 - 7 | 1294 | 132 | ||
| NM_164357.2 | NP_722595.1 | 6 - 7 | 1381 | 132 | ||
| NM_164358.2 | NP_722596.1 | 6 - 7 | 1354 | 132 | ||
| NM_164359.2 | NP_722597.1 | 6 - 7 | 1506 | 132 | ||
| NM_164360.3 | NP_722598.1 | 6 - 7 | 1430 | 132 | ||
| NM_164361.2 | NP_722599.1 | 7 - 8 | 1407 | 132 | ||
| NM_164362.3 | NP_722600.1 | 5 - 6 | 1194 | 132 | ||
| NM_205880.1 | NP_995602.1 | 7 - 8 | 1388 | 132 | ||
| GenBank mRNA | AY051918.1 | - | 6 - 7 | 1290 | 132 | |
| BT099838.1 | - | 5 - 6 | 1180 | 132 | ||
More Information
Set Membership: |
|

Sam-S
Genomic Map