Assay Details
| Gene Symbol: Eaat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169373.2 | NP_001162844.1 | 5 - 6 | 2348 | 66 | |
| NM_001169374.2 | NP_001162845.1 | 7 - 8 | 2119 | 66 | ||
| NM_058079.3 | NP_477427.1 | 7 - 8 | 2007 | 66 | ||
More Information
Set Membership: |
|

Eaat2
Genomic Map