Assay Details
| Gene Symbol: gcm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135458.5 | NP_609302.1 | 2 - 3 | 1091 | 66 | |
| GenBank mRNA | AF184664.1 | - | 2 - 3 | 1115 | 66 | |
| BT022152.1 | - | 2 - 3 | 1004 | 66 | ||
More Information
Set Membership: |
|

gcm2
Genomic Map