Assay Details
| Gene Symbol: IFT52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135201.2 | NP_609045.2 | 1 - 2 | 186 | 72 | |
| GenBank mRNA | BT120082.1 | - | 1 - 2 | 186 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IFT52| Gene Symbol: IFT52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135201.2 | NP_609045.2 | 1 - 2 | 186 | 72 | |
| GenBank mRNA | BT120082.1 | - | 1 - 2 | 186 | 72 | |
Set Membership: |
|