Assay Details
| Gene Symbol: CCDC53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135334.5 | NP_609178.1 | 2 - 3 | 532 | 92 | |
| GenBank mRNA | AY071457.1 | - | 2 - 3 | 523 | 92 | |
More Information
Set Membership: |
|

CCDC53
Genomic Map