Assay Details
| Gene Symbol: mRpL51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135395.3 | NP_609239.1 | 1 | 574 | 65 | |
| GenBank mRNA | AY051652.1 | - | 1 | 521 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mRpL51| Gene Symbol: mRpL51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135395.3 | NP_609239.1 | 1 | 574 | 65 | |
| GenBank mRNA | AY051652.1 | - | 1 | 521 | 65 | |
Set Membership: |
|