Assay Details
| Gene Symbol: sxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078896.3 | NP_523620.1 | 3 - 4 | 445 | 113 | |
| NM_165426.2 | NP_724406.1 | 3 - 4 | 504 | 113 | ||
| NM_165427.2 | NP_724407.1 | 2 - 3 | 565 | 113 | ||
| GenBank mRNA | AF217788.1 | - | 3 - 4 | 490 | 113 | |
| BT032865.1 | - | 2 - 3 | 546 | 113 | ||
More Information
Set Membership: |
|

sxc
Genomic Map