Assay Details
| Gene Symbol: sxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078896.3 | NP_523620.1 | 4 - 5 | 640 | 98 | |
| NM_165426.2 | NP_724406.1 | 4 - 5 | 699 | 98 | ||
| NM_165427.2 | NP_724407.1 | 3 - 4 | 760 | 98 | ||
| GenBank mRNA | AF217788.1 | - | 4 - 5 | 685 | 98 | |
| BT032865.1 | - | 3 - 4 | 741 | 98 | ||
More Information
Set Membership: |
|

sxc
Genomic Map