Assay Details
| Gene Symbol: sxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078896.3 | NP_523620.1 | 5 - 6 | 709 | 64 | |
| NM_165426.2 | NP_724406.1 | 5 - 6 | 768 | 64 | ||
| NM_165427.2 | NP_724407.1 | 4 - 5 | 829 | 64 | ||
| GenBank mRNA | AF217788.1 | - | 5 - 6 | 754 | 64 | |
| BT032865.1 | - | 4 - 5 | 810 | 64 | ||
More Information
Set Membership: |
|

sxc
Genomic Map