Assay Details
| Gene Symbol: sxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078896.3 | NP_523620.1 | 7 - 8 | 1098 | 66 | |
| NM_165426.2 | NP_724406.1 | 7 - 8 | 1157 | 66 | ||
| NM_165427.2 | NP_724407.1 | 6 - 7 | 1218 | 66 | ||
More Information
Set Membership: |
|

sxc
Genomic Map