Assay Details
| Gene Symbol: sxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078896.3 | NP_523620.1 | 8 - 9 | 2826 | 93 | |
| NM_165426.2 | NP_724406.1 | 8 - 9 | 2885 | 93 | ||
| NM_165427.2 | NP_724407.1 | 7 - 8 | 2946 | 93 | ||
| GenBank mRNA | AF217788.1 | - | 8 - 9 | 2871 | 93 | |
| BT032865.1 | - | 7 - 8 | 2927 | 93 | ||
More Information
Set Membership: |
|

sxc
Genomic Map