Assay Details
| Gene Symbol: Dgk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103749.2 | NP_001097219.1 | 8 - 9 | 1462 | 55 | |
| NM_001103750.3 | NP_001097220.1 | 7 - 8 | 1153 | 55 | ||
| NM_001273863.1 | NP_001260792.1 | 7 - 8 | 1153 | 55 | ||
| NM_001316408.1 | NP_001303337.1 | 7 - 8 | 1153 | 55 | ||
| NM_165568.3 | NP_724619.2 | 9 - 10 | 1543 | 55 | ||
More Information
Set Membership: |
|

Dgk
Genomic Map